(±)-3,5-Dimethyl-1-hexyn-3-ol ≥98.0%
Supplier: TCI America
D0737-500ML
D0737-25ML
TCD0737-500MLEA
114
USD
TCD0737-500ML
TCD0737-025ML
(±)-3,5-Dimethyl-1-hexyn-3-ol ≥98.0%
(±)-3,5-Dimethyl-1-hexyn-3-ol
CAS Number: 107-54-0
MDL Number: MFCD00008941
Molecular Formula: C8H14O
Molecular Weight: 126.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): 57
Specific Gravity (20/20): 0.86
Formula:
C₈H₁₄O Boiling Pt: 150 °C Density: 0.86 (20 °C) Flash Pt: 57 °C |
MDL Number:
MFCD00008941 CAS Number: 107-54-0 EINECS: 203-500-9 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...