Zinc dimethyldithiocarbamate ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Dimethyldithiocarbamic Acid Zinc Salt, Ziram
D0717-500G
D0717-25G
TCD0717-500GEA
62.05
USD
TCD0717-500G
TCD0717-025G
Zinc dimethyldithiocarbamate ≥97.0% (by titrimetric analysis)
Zinc dimethyldithiocarbamate
CAS Number: 137-30-4
MDL Number: MFCD00064797
Molecular Formula: C3H7NS2
Molecular Weight: 305.80
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 257
Flash Point (°C): 264
Product Categories: Bioscience, Fungicide Ingredients, Analytical Science, Estradiol, etc. (Environmental Endocrine Disruptors)
Other Product Names / Synonyms: Dimethyldithiocarbamic Acid Zinc Salt, Ziram
Formula:
C₃H₇NS₂ MW: 305.80 g/mol Melting Pt: 257 °C Flash Pt: 264 °C |
MDL Number:
MFCD00064797 CAS Number: 137-30-4 |
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...