Sodium dimethyldithiocarbamate dihydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Sodium Dimethyldithiocarbamate Dihydrate, Dimethyldithiocarbamic acid sodium salt dihydrate
D0716-25G
D0716-500G
TCD0716-025GEA
33
USD
TCD0716-025G
TCD0716-500G
Sodium dimethyldithiocarbamate dihydrate ≥98.0% (by titrimetric analysis)
Sodium dimethyldithiocarbamate dihydrate
CAS Number: 128-04-1
MDL Number: MFCD00150729
Molecular Formula: C3H7NS2
Molecular Weight: 143.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: Dimethyldithiocarbamic Acid Sodium Salt
Formula:
C₃H₁₀NNaO₂S₂ MW: 179.24 g/mol |
MDL Number:
MFCD00150729 CAS Number: 72140-17-1 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...