Dimethylammonium dimethyldithiocarbamate ≥95.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Dimethyldithiocarbamic Acid Dimethylammonium Salt
D0715-25G
D0715-5G
TCD0715-025GEA
234
USD
TCD0715-025G
TCD0715-5G
Dimethylammonium dimethyldithiocarbamate ≥95.0% (by titrimetric analysis)
Dimethylammonium dimethyldithiocarbamate
CAS Number: 598-64-1
MDL Number: MFCD00067267
Molecular Formula: C5H14N2S2
Molecular Weight: 166.30
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 132
Other Product Names / Synonyms: Dimethyldithiocarbamic Acid Dimethylammonium Salt
Formula:
C₅H₁₄N₂S₂ MW: 166.30 g/mol Melting Pt: 132 °C |
MDL Number:
MFCD00067267 CAS Number: 598-64-1 |
Specification Test Results
Purity/Analysis Method | >95.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...