2,4-Dimethyl-1,3-dioxane 98,0
Supplier: TCI America
D0708-25ML
D0708-10ML
TCD0708-025MLEA
278
USD
TCD0708-025ML
TCD0708-010ML
2,4-Dimethyl-1,3-dioxane 98,0
2,4-Dimethyl-1,3-dioxane
CAS Number: 766-20-1
MDL Number: MFCD00023821
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method:
Formula:
C₆H₁₂O₂ MW: 116.16 g/mol Boiling Pt: 118 °C Density: 0.94 (20 °C) |
MDL Number:
MFCD00023821 CAS Number: 766-20-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...