1,4-Dimethylcyclohexane ≥98.0%

Supplier: TCI America
Danger

Synonyms: Hexahydro-p-xylene

D0700-500ML D0700-25ML
TCD0700-500MLEA 505 USD
TCD0700-500ML TCD0700-025ML
1,4-Dimethylcyclohexane ≥98.0%
1,4-Dimethylcyclohexane

CAS Number: 589-90-2
MDL Number: MFCD00001508
Molecular Formula: C8H16
Molecular Weight: 112.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 10
Specific Gravity (20/20): 0.77


Other Product Names / Synonyms: Hexahydro-p-xylene

Formula: C₈H₁₆
MW: 112.22 g/mol
Boiling Pt: 120 °C (1013 hPa)
Melting Pt: -87 °C
Density: 0.77 (20 °C)
Flash Pt: 10 °C
MDL Number: MFCD00001508
CAS Number: 589-90-2
EINECS: 209-663-2
UN: 2263
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6b61e1e590b3a20f94b2412109c27fccb70f271&vid=fbb2977156f52e3cd8ec484abb1370c9be4fd492&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR