3,4-Dimethoxyphenethylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(3,4-Dimethoxyphenyl)ethylamine

D0678-25G D0678-500G
TCD0678-025GEA 73.86 USD
TCD0678-025G TCD0678-500G
3,4-Dimethoxyphenethylamine ≥98.0% (by GC, titration analysis)
3,4-Dimethoxyphenethylamine

CAS Number: 120-20-7
MDL Number: MFCD00008188
Molecular Formula: C10H15NO2
Molecular Weight: 181.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 156
Flash Point (°C): 130
Freezing point (°C): 11
Specific Gravity (20/20): 1.10


Other Product Names / Synonyms: 2-(3,4-Dimethoxyphenyl)ethylamine

Formula: C₁₀H₁₅NO₂
MW: 181.23 g/mol
Boiling Pt: 156 °C
Density: 1.10 (20 °C)
Flash Pt: 130 °C
MDL Number: MFCD00008188
CAS Number: 120-20-7
EINECS: 204-376-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efec8c3855d94e4945df73d56f00532e5db15350&vid=efec8c3855d94e4945df73d56f00532e5db15350&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR