1-(Dimethylamino)-2-propanol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
CAS Number: 108-16-7
MDL Number: MFCD00004532
Molecular Formula: C5H13NO
Molecular Weight: 103.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 125
Melting point (°C): -85
Flash Point (°C): 32
Specific Gravity (20/20): 0.85
Other Product Names / Synonyms: N,N-Dimethylisopropanolamine
Formula:
C₅H₁₃NO Boiling Pt: 125 °C Melting Pt: –85 °C Density: 0.85 (20 °C) Flash Pt: 32 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004532 CAS Number: 108-16-7 EINECS: 203-556-4 UN: 2733 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...