4-(4-Dimethylaminophenylazo)phenylarsonic acid hydrochloride ≥95.0% (by titrimetric analysis)

Supplier: TCI America

D0659-25G D0659-1G
TCD0659-025GEA 557.6 USD
TCD0659-025G TCD0659-001G
4-(4-Dimethylaminophenylazo)phenylarsonic acid hydrochloride ≥95.0% (by titrimetric analysis)
4-(4-Dimethylaminophenylazo)phenylarsonic acid hydrochloride

CAS Number: 73688-85-4
MDL Number: MFCD00012589
Molecular Formula: C14H16AsN3O3
Molecular Weight: 385.68
Purity/Analysis Method: >95.0% (T)
Form: Crystal


Other Product Names / Synonyms: 4-(4-Dimethylaminophenylazo)benzenearsonic Acid Hydrochloride

Formula: C₁₄H₁₆AsN₃O₃
MDL Number: MFCD00012589
CAS Number: 73688-85-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9268ce2be66f76bfa3b328e072fa56d9fd1fca72&vid=9268ce2be66f76bfa3b328e072fa56d9fd1fca72&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR