3,4-Dimethoxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D0631-25G D0631-500G
TCD0631-025GEA 74 USD
TCD0631-025G TCD0631-500G
3,4-Dimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
3,4-Dimethoxybenzoic acid

CAS Number: 93-07-2
MDL Number: MFCD00002500
Molecular Formula: C9H10O4
Molecular Weight: 182.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 182


Other Product Names / Synonyms: Protocatechuic Acid Dimethyl Ether, Veratric Acid

Formula: C₉H₁₀O₄
Boiling Pt: 303 °C (1013 hPa)
Melting Pt: 182 °C
Density: 1.436 g/cm³ (–100 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002500
CAS Number: 93-07-2
EINECS: 202-215-7
Merck Index: 12,10086
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9dc9dd80e7df8c4367823752042bc49574ea04d&vid=512439d0069359721857146ff5795d178bb39e64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR