DL-Adrenaline hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
Racepinephrine Hydrochloride, DL-Epinephrine Hydrochloride
A0909-25G
A0909-5G
TCA0909-025GEA
355
USD
TCA0909-025G
TCA0909-5G
DL-Adrenaline hydrochloride ≥98.0%
DL-Adrenaline hydrochloride
CAS Number: 329-63-5
MDL Number: MFCD00050562
Molecular Formula: C9H13NO3
Molecular Weight: 219.67
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 157
Product Categories: Bioscience, Phenylethylamines
Other Product Names / Synonyms: DL-Epinephrine Hydrochloride
Formula:
C₉H₁₃NO₃ MW: 219.67 g/mol Melting Pt: 157 °C |
MDL Number:
MFCD00050562 CAS Number: 329-63-5 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...