3,5-Diiodo-L-tyrosine dihydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: H-Tyr(3,5-DiI)-OH·2H₂O, (S)-2-Amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid dihydrate

D0612-25G D0612-5G
TCD0612-025GEA 68.28 USD
TCD0612-025G TCD0612-5G
3,5-Diiodo-L-tyrosine dihydrate ≥98.0% (by titrimetric analysis)
3,5-Diiodo-L-tyrosine dihydrate

CAS Number: 18835-59-1
MDL Number: MFCD00016542
Molecular Formula: C9H9I2NO3
Molecular Weight: 432.98
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 213
Specific rotation [a]20/D: 1.2 deg (C=5, 1mol/L HCl)

Formula: C₉H₉I₂NO₃·2H₂O
MW: 469.01 g/mol
Melting Pt: 213 °C
MDL Number: MFCD00016542
CAS Number: 18835-59-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea2758371e6ad1e96805ddd1c42ce6438dac37c5&vid=bea71cf8f54f1ff713a3ec5628068648e924282d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR