Diiodomethane ≥98.0% stabilized
Supplier: TCI America
Synonyms:
Methylene iodide
D0610-25G
D0610-500G
D0610-100G
TCD0610-025GEA
43.74
USD
TCD0610-025G
TCD0610-500G
TCD0610-100G
Diiodomethane ≥98.0% stabilized
Diiodomethane
(stabilized with Copper chip)
CAS Number: 75-11-6
MDL Number: MFCD00001079
Molecular Formula: CH2I2
Molecular Weight: 267.84
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 181
Flash Point (°C): 110
Freezing point (°C): 6
Specific Gravity (20/20): 3.33
Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination, Halogenation
Other Product Names / Synonyms: Methylene Iodide
Formula:
CH₂I₂ MW: 267.84 g/mol Boiling Pt: 181 °C Melting Pt: 6 °C Density: 3.33 (20 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001079 CAS Number: 75-11-6 EINECS: 200-841-5 UN: 2810 ADR: 6.1,III Merck Index: 12,06143 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...