Diethylstilbestrol cis- and trans-mixture ≥98.0%

Supplier: TCI America

Synonyms: (E)-3,4-Bis(4-hydroxyphenyl)-3-hexene, Stilbestrol, DES, (E)-4,4'-(Hex-3-ene-3,4-diyl)diphenol

D0526-5G D0526-1G
TCD0526-005GEA 118.2 USD
TCD0526-005G TCD0526-001G
Diethylstilbestrol cis- and trans-mixture ≥98.0%
Diethylstilbestrol

CAS Number: 6898-97-1
MDL Number: MFCD00002373
Molecular Formula: C18H20O2
Molecular Weight: 268.36
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 169


Other Product Names / Synonyms: Stilbestrol

Formula: C₁₈H₂₀O₂
MW: 268.356 g/mol
Melting Pt: 169 °C
MDL Number: MFCD00002373
CAS Number: 56-53-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4047e94decd894a5c09da72598a5d0b52b8fcfcf&vid=b8ef7219e21265e1977a7192a76e2d7c366db75c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR