N,N-Diethylcyclohexylamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
D0484-25ML
D0484-500ML
TCD0484-025MLEA
41.7
USD
TCD0484-025ML
TCD0484-500ML
N,N-Diethylcyclohexylamine ≥98.0% (by GC, titration analysis)
N,N-Diethylcyclohexylamine
CAS Number: 91-65-6
MDL Number: MFCD00003847
Molecular Formula: C10H21N
Molecular Weight: 155.29
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 193
Flash Point (°C): 57
Specific Gravity (20/20): 0.84
Formula:
C₁₀H₂₁N MW: 155.2804 g/mol Boiling Pt: 193 °C Density: 0.84 (20 °C) Flash Pt: 57 °C |
MDL Number:
MFCD00003847 CAS Number: 91-65-6 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...