Dichloro(diphenyl)silane ≥98.0% for synthesis

Supplier: TCI America

Synonyms: Diphenyldichlorosilane, Dichlorodiphenylsilane

D0362-500G D0362-25G D0362-100G
TCD0362-500GEA 179 USD
TCD0362-500G TCD0362-025G TCD0362-100G
Dichloro(diphenyl)silane ≥98.0% for synthesis
Dichloro(diphenyl)silane

CAS Number: 80-10-4
MDL Number: MFCD00000489
Molecular Formula: C12H10Cl2Si
Molecular Weight: 253.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 303
Flash Point (°C): 157
Specific Gravity (20/20): 1.22

Formula: C₁₂H₁₀Cl₂Si
Boiling Pt: 303 °C
Density: 1.22 (20 °C)
Flash Pt: 157 °C
MDL Number: MFCD00000489
CAS Number: 80-10-4
EINECS: 201-251-0
UN: 1769
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a910ad4362ffe1934cc4c7b08f0d1d3499038f1f&vid=a910ad4362ffe1934cc4c7b08f0d1d3499038f1f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR