2,4-Dichlorobenzophenone ≥97.0%
Supplier: TCI America
D0342-5G
D0342-25G
TCD0342-5GEA
64
USD
TCD0342-5G
TCD0342-25G
2,4-Dichlorobenzophenone ≥97.0%
2,4-Dichlorobenzophenone
CAS Number: 19811-05-3
MDL Number: MFCD00018374
Molecular Formula: C13H8Cl2O
Molecular Weight: 251.11
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 47
Formula:
C₁₃H₈Cl₂O Melting Pt: 47 °C |
MDL Number:
MFCD00018374 CAS Number: 19811-05-3 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...