Dibutyltin maleate (so called) (so called)

Supplier: TCI America

D0304-25G D0304-500G
TCD0304-025GEA 33 USD
TCD0304-025G TCD0304-500G
Dibutyltin maleate (so called) (so called)
Dibutyltin maleate (so called)

[for PVC stabilizer]
CAS Number: 78-04-6
MDL Number: MFCD00014120
Molecular Formula: C12H20O4Sn
Molecular Weight: 347.00
Form: Clear Liquid
Color: Slightly Pale Yellow
Flash Point (°C): 132
Specific Gravity (20/20): 1.28

Formula: C₁₂H₂₀O₄Sn
MW: 347.00 g/mol
Density: 1.28 (20 °C)
Flash Pt: 132 °C
MDL Number: MFCD00014120
CAS Number: 78-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR