Dipentamethylenethiuram tetrasulfide

Supplier: TCI America

Synonyms: Bis(pentamethylenethiocarbamoyl) Tetrasulphide, Tetrasulphanediylbis(piperidin-1-ylmethanethione)

D0279-25G D0279-500G
TCD0279-025GEA 29.04 USD
TCD0279-025G TCD0279-500G
Dipentamethylenethiuram tetrasulfide
Dipentamethylenethiuram tetrasulfide

[Vulcanization Accelerator]
CAS Number: 120-54-7
MDL Number: MFCD00047474
Molecular Formula: C12H20N2S6
Molecular Weight: 384.67
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 120


Other Product Names / Synonyms: Bis(pentamethylenethiocarbamoyl) Tetrasulfide

Formula: C₁₂H₂₀N₂S₆
MW: 384.67 g/mol
Melting Pt: 120 °C
MDL Number: MFCD00047474
CAS Number: 120-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR