2,5-Dimethylpyrrole ≥98.0%
Supplier: TCI America
Synonyms:
2,5-Dimethyl-1H-pyrrole
D0260-25ML
D0260-5ML
TCD0260-025MLEA
114
USD
TCD0260-025ML
TCD0260-5ML
2,5-Dimethylpyrrole ≥98.0%
2,5-Dimethylpyrrole
CAS Number: 625-84-3
MDL Number: MFCD00005223
Molecular Formula: C6H9N
Molecular Weight: 95.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 169
Flash Point (°C): 54
Specific Gravity (20/20): 0.94
Formula:
C₆H₉N MW: 95.1424 g/mol Boiling Pt: 169 °C Density: 0.94 (20 °C) Flash Pt: 54 °C |
MDL Number:
MFCD00005223 CAS Number: 625-84-3 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...