Zinc(II) dibutyldithiocarbamate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D0227-500G D0227-25G
TCD0227-500GEA 190 USD
TCD0227-500G TCD0227-025G
Zinc(II) dibutyldithiocarbamate ≥98.0% (by titrimetric analysis)
Zinc(II) dibutyldithiocarbamate

CAS Number: 136-23-2
MDL Number: MFCD00067274
Molecular Formula: C9H19NS2
Molecular Weight: 474.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 109
Flash Point (°C): 181


Other Product Names / Synonyms: Dibutyldithiocarbamic Acid Zinc(II) Salt

Formula: C₉H₁₉NS₂
Melting Pt: 109 °C
Flash Pt: 181 °C
MDL Number: MFCD00067274
CAS Number: 136-23-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1fa60cb9cd53042b748c045b40c1a9558c477b1&vid=e1fa60cb9cd53042b748c045b40c1a9558c477b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR