N-(2-Aminoethyl)morpholine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4-(2-Aminoethyl)morpholine, 2-Morpholinoethanamine
A0883-100G
A0883-25G
A0883-500G
TCA0883-100GEA
94
USD
TCA0883-100G
TCA0883-25G
TCA0883-500G
N-(2-Aminoethyl)morpholine ≥98.0% (by GC, titration analysis)
N-(2-Aminoethyl)morpholine
CAS Number: 2038-03-1
MDL Number: MFCD00006182
Molecular Formula: C6H14N2O
Molecular Weight: 130.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 205
Flash Point (°C): 175
Freezing point (°C): 25
Other Product Names / Synonyms: 2-Morpholinoethylamine
Formula:
C₆H₁₄N₂O MW: 130.19 g/mol Boiling Pt: 205 °C Flash Pt: 175 °C |
MDL Number:
MFCD00006182 CAS Number: 2038-03-1 EINECS: 218-011-6 UN: 2923 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...