(±)-2,3-Dibromo-1-propanol ≥95.0%
Supplier: TCI America
|
Danger
|
Synonyms:
2,3-Dibromo-1-propanol, Glycerol alpha,beta-Dibromohydrin, alpha,beta-Dibromohydrin
D0204-25G
D0204-500G
TCD0204-025GEA
28
USD
TCD0204-025G
TCD0204-500G
(±)-2,3-Dibromo-1-propanol ≥95.0%
(±)-2,3-Dibromo-1-propanol
CAS Number: 96-13-9
MDL Number: MFCD00004699
Molecular Formula: C3H6Br2O
Molecular Weight: 217.89
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 99
Melting point (°C): 9
Flash Point (°C): 110
Specific Gravity (20/20): 2.14
Other Product Names / Synonyms: alpha,beta-Dibromohydrin, Glycerol alpha,beta-Dibromohydrin
Formula:
HOCH₂BrCHCH₂Br MW: 217.89 g/mol Boiling Pt: 99 °C Melting Pt: 9 °C Density: 2.14 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00004699 CAS Number: 96-13-9 EINECS: 202-480-9 |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...