Phenoxybenzamine hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
Dibenzyline Hydrochloride, N-(2-Chloroethyl)-N-(1-methyl-2-phenoxyethyl)benzylamine Hydrochloride, N-Phenoxyisopropyl-N-benzyl-2-chloroethylamine Hydrochloride, N-Benzyl-N-(2-chloroethyl)-1-phenoxypropan-2-amine hydrochloride
CAS Number: 63-92-3
MDL Number: MFCD00055152
Molecular Formula: C18H22ClNO
Molecular Weight: 340.29
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 139
Other Product Names / Synonyms: N-(2-Chloroethyl)-N-(1-methyl-2-phenoxyethyl)benzylamine Hydrochloride, Dibenzyline Hydrochloride, N-Phenoxyisopropyl-N-benzyl-2-chloroethylamine Hydrochloride
Formula:
C₁₈H₂₂ClNO MW: 340.29 g/mol Melting Pt: 139 °C |
MDL Number:
MFCD00055152 CAS Number: 63-92-3 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...