4-Diazo-N,N-dimethylaniline chloride zinc chloride hydrate ≥95.0%
Supplier: TCI America
Synonyms:
N,N-Dimethyl-1,4-phenylenediamine Diazonium Chloride Zinc Chloride
D0140-500G
D0140-25G
TCD0140-500GEA
147
USD
TCD0140-500G
TCD0140-025G
4-Diazo-N,N-dimethylaniline chloride zinc chloride hydrate ≥95.0%
4-Diazo-N,N-dimethylaniline chloride zinc chloride hydrate
CAS Number: 6087-56-5
MDL Number: MFCD00031685
Molecular Formula: C8H10ClN3
Molecular Weight: 319.92
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Other Product Names / Synonyms: N,N-Dimethyl-1,4-phenylenediamine Diazonium Chloride Zinc Chloride
Formula:
C₈H₁₀ClN₃ MW: 319.92 g/mol |
MDL Number:
MFCD00031685 CAS Number: 6087-56-5 |
|
Specification Test Results
Purity/Analysis Method | >95.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...