m-Xylylenediamine ≥99.0%

Supplier: TCI America

Synonyms: 1,3-Benzenedimethanamine

D0127-25G D0127-500G
TCD0127-025GEA 25 USD
TCD0127-025G TCD0127-500G
m-Xylylenediamine ≥99.0%
m-Xylylenediamine

CAS Number: 1477-55-0
MDL Number: MFCD00008119
Molecular Formula: C8H12N2
Molecular Weight: 136.20
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 248
Flash Point (°C): 134
Freezing point (°C): 14
Specific Gravity (20/20): 1.05


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: alpha,alpha'-Diamino-m-xylene, m-Xylene Diamine, 3-(Aminomethyl)benzylamine

Formula: C₈H₁₂N₂
MW: 136.1943 g/mol
Boiling Pt: 248 °C
Density: 1.05 (20 °C)
Flash Pt: 134 °C
MDL Number: MFCD00008119
CAS Number: 1477-55-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07e2b0d4bb38ba40a063281ea56318e8c7ade545&vid=07e2b0d4bb38ba40a063281ea56318e8c7ade545&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR