DL-2,3-Diaminopropionic acid hydrobromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3-Amino-DL-alanine Hydrobromide, 2,3-Diaminopropanoic acid hydrobromide, H-DL-Dapa-OH·HBr

D0115-25G D0115-5G
TCD0115-025GEA 478.1 USD
TCD0115-025G TCD0115-005G
DL-2,3-Diaminopropionic acid hydrobromide ≥98.0% (by titrimetric analysis)
DL-2,3-Diaminopropionic acid hydrobromide

CAS Number: 18635-45-5
MDL Number: MFCD00012883
Molecular Formula: C3H8N2O2
Molecular Weight: 185.02
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 230


Other Product Names / Synonyms: 3-Amino-DL-alanine Hydrobromide

Formula: C₃H₈N₂O₂
MW: 185.02 g/mol
Melting Pt: 230 °C
MDL Number: MFCD00012883
CAS Number: 18635-45-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8d2d31718d5a86ca2aaab633a838e78a77d48b7&vid=f8d2d31718d5a86ca2aaab633a838e78a77d48b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR