2,4-Diaminoanisole dihydrochloride ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-Methoxybenzene-1,3-diamine dihydrochloride

D0074-5G D0074-25G
TCD0074-5GEA 124.5 USD
TCD0074-5G TCD0074-025G
2,4-Diaminoanisole dihydrochloride ≥99.0% (by titrimetric analysis)
2,4-Diaminoanisole dihydrochloride

CAS Number: 614-94-8
MDL Number: MFCD00054337
Molecular Formula: C7H10N2O
Molecular Weight: 211.09
Purity/Analysis Method: >99.0% (T)
Form: Crystal


Other Product Names / Synonyms: 4-Methoxy-1,3-phenylenediamine Dihydrochloride

Formula: C₇H₁₂Cl₂N₂O
MDL Number: MFCD00054337
CAS Number: 614-94-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dbae50b4d1f9c6c6dee40434c924168d5ba9442&vid=5dbae50b4d1f9c6c6dee40434c924168d5ba9442&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR