α-Bromo-2-chloro-5-fluorotoluene ≥96.0%

Supplier: TCI America

C2359-1G C2359-5G
TCC2359-1GEA 46 USD
TCC2359-1G TCC2359-5G
α-Bromo-2-chloro-5-fluorotoluene ≥96.0%
α-Bromo-2-chloro-5-fluorotoluene

CAS Number: 81778-09-8
MDL Number: MFCD01631419
Molecular Formula: C7H5BrClF
Molecular Weight: 223.47
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 66
Flash Point (°C): 18
Specific Gravity (20/20): 1.67


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-(Bromomethyl)-2-chloro-5-fluorobenzene

Formula: C₇H₅BrClF
MW: 223.47 g/mol
Boiling Pt: 66 °C
Density: 1.67 (20 °C)
Flash Pt: 18 °C
MDL Number: MFCD01631419
CAS Number: 81778-09-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2267f11d57aeb0a41c65ad48274b53b210375547&vid=2267f11d57aeb0a41c65ad48274b53b210375547&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR