Cetirizine dihydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
Cetirizine 2HCl, 2-(2-(4-((4-Chlorophenyl)(phenyl)methyl)piperazin-1-yl)ethoxy)acetic acid dihydrochloride
C2316-25G
C2316-5G
TCC2316-25GEA
290
USD
TCC2316-25G
TCC2316-5G
Cetirizine dihydrochloride ≥98.0%
Cetirizine dihydrochloride
CAS Number: 83881-52-1
MDL Number: MFCD00941428
Molecular Formula: C21H25ClN2O3
Molecular Weight: 461.81
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 225
Formula:
C₂₁H₂₅ClN₂O₃ MW: 461.81 g/mol Melting Pt: 225 °C |
MDL Number:
MFCD00941428 CAS Number: 83881-52-1 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...