5-Chloro-2-fluoropyridine ≥96.0%
Supplier: TCI America
CAS Number: 1480-65-5
MDL Number: MFCD04112513
Molecular Formula: C5H3ClFN
Molecular Weight: 131.53
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 149
Flash Point (°C): 52
Specific Gravity (20/20): 1.33
Formula:
C₅H₃ClFN Boiling Pt: 149 °C Density: 1.33 (20 °C) Flash Pt: 52 °C |
MDL Number:
MFCD04112513 CAS Number: 1480-65-5 UN: 1992 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method | >96.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...