6-Chloro-3-methyluracil ≥98.0%
Supplier: TCI America
Synonyms:
6-Chloro-3-methylpyrimidine-2,4(1H,3H)-dione
C2300-25G
C2300-5G
TCC2300-25GEA
116.1
USD
TCC2300-25G
TCC2300-5G
6-Chloro-3-methyluracil ≥98.0%
6-Chloro-3-methyluracil
CAS Number: 4318-56-3
MDL Number: MFCD01074837
Molecular Formula: C5H5ClN2O2
Molecular Weight: 160.56
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 278
Formula:
C₅H₅ClN₂O₂ MW: 160.56 g/mol Melting Pt: 278 °C |
MDL Number:
MFCD01074837 CAS Number: 4318-56-3 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...