Clindamycin phosphate ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Clindamycin 2-Dihydrogen Phosphate, (2R,3R,4S,5R,6R)-6-((1S,2S)-2-Chloro-1-((2S,4R)-1-methyl-4-propylpyrrolidine-2-carboxamido)propyl)-4,5-dihydroxy-2-(methylthio)tetrahydro-2H-pyran-3-yl dihydrogen phosphate
CAS Number: 24729-96-2
MDL Number: MFCD07793328
Molecular Formula: C18H34ClN2O8PS
Molecular Weight: 504.96
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 114
Specific rotation [a]20/D: 122 deg (C=1, H2O)
Product Categories: Bioscience, Antibiotic Ingredients
Other Product Names / Synonyms: Clindamycin 2-Dihydrogen Phosphate
Formula:
C₁₈H₃₄ClN₂O₈PS MW: 504.96 g/mol Melting Pt: 114 °C |
MDL Number:
MFCD07793328 CAS Number: 24729-96-2 |
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...