Clindamycin hydrochloride monohydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
C2256-25G
C2256-5G
TCC2256-25GEA
410
USD
TCC2256-25G
TCC2256-5G
Clindamycin hydrochloride monohydrate ≥98.0% (by titrimetric analysis)
Clindamycin hydrochloride monohydrate
CAS Number: 58207-19-5
MDL Number: MFCD06411091
Molecular Formula: C18H33ClN2O5S
Molecular Weight: 461.44
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 143
Specific rotation [a]20/D: 143 deg (C=2, H2O)
Formula:
C₁₈H₃₃ClN₂O₅S MW: 461.44 g/mol Melting Pt: 143 °C |
MDL Number:
MFCD06411091 CAS Number: 58207-19-5 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...