Cloxacillin sodium salt monohydrate ≥98.0%

Supplier: TCI America

C2244-5G C2244-1G
TCC2244-5GEA 145 USD
TCC2244-5G TCC2244-1G
Cloxacillin sodium salt monohydrate ≥98.0%
Cloxacillin sodium salt monohydrate

CAS Number: 7081-44-9
MDL Number: MFCD00150735
Molecular Formula: C19H18ClN3O5S
Molecular Weight: 457.86
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 164 deg (C=1, H2O)

Formula: C₁₉H₁₈ClN₃O₅S
MDL Number: MFCD00150735
CAS Number: 7081-44-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae60c160e45275b592915b8ee11b92d2a65d92e8&vid=ae60c160e45275b592915b8ee11b92d2a65d92e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR