Cefazolin sodium salt ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Sodium (6R,7R)-7-(2-(1H-tetrazol-1-yl)acetamido)-3-(((5-methyl-1,3,4-thiadiazol-2-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

C2242-5G C2242-25G
TCC2242-5GEA 109.85 USD
TCC2242-5G TCC2242-25G
Cefazolin sodium salt ≥98.0% (by HPLC, titration analysis)
Cefazolin sodium salt

CAS Number: 27164-46-1
MDL Number: MFCD00056883
Molecular Formula: C14H14N8O4S3
Molecular Weight: 476.48
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 190
Specific rotation [a]20/D: -20 deg (C=10, H2O)
Storage Temperature: 0-10°C

Formula: C₁₄H₁₄N₈NaO₄S₃
MW: 476.48 g/mol
Melting Pt: 190 °C
MDL Number: MFCD00056883
CAS Number: 27164-46-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2e65161075519c88224e537dff5af93067cc101&vid=f2e65161075519c88224e537dff5af93067cc101&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR