ɑ-Amylcinnamaldehyde ≥90.0%

Supplier: TCI America

Synonyms: Amylcinnamal, 2-Amyl-3-phenylacrylaldehyde, 2-Benzylideneheptanal, ɑ-Pentylcinnamaldehyde, 2-Amyl-3-phenyl-2-propenal, Jasminal

A0851-500ML A0851-25ML
TCA0851-500MLEA 103 USD
TCA0851-500ML TCA0851-025ML
ɑ-Amylcinnamaldehyde ≥90.0%
alpha-Amylcinnamaldehyde

CAS Number: 122-40-7
MDL Number: MFCD00006988
Molecular Formula: C14H18O
Molecular Weight: 202.30
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 175
Flash Point (°C): 113
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: Jasminal, alpha-Pentylcinnamaldehyde, 2-Amyl-3-phenylacrylaldehyde, 2-Amyl-3-phenyl-2-propenal

Formula: C₁₄H₁₈O
MW: 202.30 g/mol
Boiling Pt: 175 °C
Density: 0.97 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00006988
CAS Number: 122-40-7

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR