2-Chloroethyl benzenesulfonate ≥98.0%

Supplier: TCI America

Synonyms: Benzenesulphonic Acid 2-Chloroethyl Ester

C2230-25G C2230-5G
TCC2230-25GEA 368 USD
TCC2230-25G TCC2230-5G
2-Chloroethyl benzenesulfonate ≥98.0%
2-Chloroethyl benzenesulfonate

CAS Number: 16670-48-7
Molecular Formula: C8H9ClO3S
Molecular Weight: 220.67
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 188
Specific Gravity (20/20): 1.35


Other Product Names / Synonyms: Benzenesulfonic Acid 2-Chloroethyl Ester

Formula: C₈H₉ClO₃S
Boiling Pt: 188 °C
Density: 1.35 (20 °C)
CAS Number: 16670-48-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71653fce039c79cfaab7826f787dbeeff0aca2d4&vid=71653fce039c79cfaab7826f787dbeeff0aca2d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR