Cefotaxime sodium salt ≥96.0%

Supplier: TCI America

Synonyms: Sodium (6R,7R)-3-(acetoxymethyl)-7-((Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)acetamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, Cefotaxime sodium

C2224-5G C2224-25G
TCC2224-5GEA 133.58 USD
TCC2224-5G TCC2224-25G
Cefotaxime sodium salt ≥96.0%
Cefotaxime sodium salt

CAS Number: 64485-93-4
MDL Number: MFCD00079073
Molecular Formula: C16H17N5O7S2
Molecular Weight: 477.44
Purity/Analysis Method: >96.0% (HPLC)
Form: Crystal
Melting point (°C): 163
Specific rotation [a]20/D: 61 deg (C=1, H2O)
Storage Temperature: 0-10°C

Formula: C₁₆H₁₇N₅O₇S₂
MW: 477.44 g/mol
Melting Pt: 163 °C
MDL Number: MFCD00079073
CAS Number: 64485-93-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba78c8b74ab10e75b5b5eb855ba99dce2c72de41&vid=ba78c8b74ab10e75b5b5eb855ba99dce2c72de41&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR