N-Carbobenzoxy-D-tyrosine ≥98.0%

Supplier: TCI America

Synonyms: (R)-2-(((Benzyloxy)carbonyl)amino)-3-(4-hydroxyphenyl)propanoic acid, N-Cbz-D-tyrosine, Z-D-Tyr-OH

C2124-1G C2124-5G
TCC2124-1GEA 47 USD
TCC2124-1G TCC2124-5G
N-Carbobenzoxy-D-tyrosine ≥98.0%
N-Carbobenzoxy-D-tyrosine

CAS Number: 64205-12-5
MDL Number: MFCD00063031
Molecular Formula: C17H17NO5
Molecular Weight: 315.33
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 99


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-D-tyrosine, Z-D-Tyr-OH

Formula: C₁₇H₁₇NO₅
MW: 315.33 g/mol
Melting Pt: 99 °C
MDL Number: MFCD00063031
CAS Number: 64205-12-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc61ea14099d7e3c761e3b71b8c91da8b12777d4&vid=fc61ea14099d7e3c761e3b71b8c91da8b12777d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR