2-Chlorophenyl acetate ≥98.0%
Supplier: TCI America
CAS Number: 4525-75-1
MDL Number: MFCD01632281
Molecular Formula: C8H7ClO2
Molecular Weight: 170.59
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 103
Melting point (°C): -20
Specific Gravity (20/20): 1.22
Other Product Names / Synonyms: Acetic Acid 2-Chlorophenyl Ester, 1-Acetoxy-2-chlorobenzene
Formula:
C₈H₇ClO₂ Boiling Pt: 103 °C Melting Pt: –20 °C Density: 1.22 (20 °C) |
MDL Number:
MFCD01632281 CAS Number: 4525-75-1 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...