3-Chloro-2-hydroxypropyl methacrylate ≥83.0% stabilised

Supplier: TCI America

Synonyms: Methacrylic Acid 3-Chloro-2-hydroxypropyl Ester

C2105-25G C2105-500G
TCC2105-25GEA 23.38 USD
TCC2105-25G TCC2105-500G
3-Chloro-2-hydroxypropyl methacrylate ≥83.0% stabilised
3-Chloro-2-hydroxypropyl methacrylate

(stabilized with HQ)
CAS Number: 13159-52-9
MDL Number: MFCD00040686
Molecular Formula: C7H11ClO3
Molecular Weight: 178.61
Purity/Analysis Method: >83.0% (GC)
Form: Clear Liquid
Boiling point (°C): 108
Flash Point (°C): 138
Specific Gravity (20/20): 1.19
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Methacrylic Acid 3-Chloro-2-hydroxypropyl Ester

Formula: C₇H₁₁ClO₃
MW: 178.61 g/mol
Boiling Pt: 108 °C
Density: 1.19 (20 °C)
Flash Pt: 138 °C
MDL Number: MFCD00040686
CAS Number: 13159-52-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51c09a1456a997b3a92831243fc5310ce874813e&vid=51c09a1456a997b3a92831243fc5310ce874813e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >83.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR