Indole-4-carbonitrile ≥98.0%

Supplier: TCI America

Synonyms: 4-Cyano-1H-indole, 1H-Indole-4-carbonitrile, 4-Cyanoindole

C2083-1G C2083-5G
TCC2083-1GEA 30.3 USD
TCC2083-1G TCC2083-5G
Indole-4-carbonitrile ≥98.0%
Indole-4-carbonitrile

CAS Number: 16136-52-0
MDL Number: MFCD00152045
Molecular Formula: C9H6N2
Molecular Weight: 142.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 120


Other Product Names / Synonyms: 4-Indolecarbonitrile

Formula: C₉H₆N₂
MW: 142.16 g/mol
Melting Pt: 120 °C
MDL Number: MFCD00152045
CAS Number: 16136-52-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9eb44cab8e6ef0272d6037f747f7d62d8a7c69f&vid=c9eb44cab8e6ef0272d6037f747f7d62d8a7c69f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR