Carboplatin ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: cis-Diammine(1,1-cyclobutanedicarboxylato)platinum(II), cis-Diamine(1,1-cyclobutanedicarboxylato)platinum(II), 1,1-Cyclobutanedicarboxylatodiammineplatinum(II)

C2043-1G C2043-100MG
TCC2043-1GEA 397 USD
TCC2043-1G TCC2043-100MG
Carboplatin ≥98.0% (by HPLC)
Carboplatin

CAS Number: 41575-94-4
Molecular Formula: C6H12N2O4Pt
Molecular Weight: 371.26
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Product Categories: Bioscience, Antitumor Ingredients
Other Product Names / Synonyms: cis-Diamine(1,1-cyclobutanedicarboxylato)platinum(II)

Formula: C₆H₁₂N₂O₄Pt
MW: 371.26 g/mol
MDL Number: MFCD00070464
CAS Number: 41575-94-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f429da9548388d6794da5d8c0983f3c095818ba&vid=2f429da9548388d6794da5d8c0983f3c095818ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR