Coenzyme Q10 ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: (all-E)-2-(3,7,11,15,19,23,27,31,35,39-Decamethyl-2,6,10,14,18,22,26,30,34,38-tetracontadecaenyl)-5,6-dimethoxy-3-methyl-2,5-cyclohexadiene-1,4-dione, Ubiquinone 10, Ubidecarenone

C1971-1G C1971-100MG
TCC1971-1GEA 308.26 USD
TCC1971-1G TCC1971-100MG
Coenzyme Q10 ≥98.0% (by HPLC)
Coenzyme Q10

CAS Number: 303-98-0
Molecular Formula: C59H90O4
Molecular Weight: 863.37
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 49
Storage Temperature: 0-10°C


Product Categories: Bioscience, Antioxidants, Vitamin Related Compounds, Terpenes (Others)
Other Product Names / Synonyms: Ubidecarenone, Ubiquinone 10

Formula: C₅₉H₉₀O₄
MW: 863.37 g/mol
Melting Pt: 49 °C
MDL Number: MFCD00042919
CAS Number: 303-98-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3dbedb1fbe2fb04855ed8ddcec0d10a89d118a28&vid=3dbedb1fbe2fb04855ed8ddcec0d10a89d118a28&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR