1-(3-Aminopropyl)-2-methylpiperidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1-(3-Aminopropyl)-2-methylhexahydropyridine, 2-Methyl-1-piperidinepropanamine, N-(3-Aminopropyl)-2-pipecoline, 3-(2-Methylpiperidin-1-yl)propan-1-amine, 1-(3-Aminopropyl)-2-methylpiperidine, 1-(3-Aminopropyl)-2-pipecoline

A0833-25ML A0833-500ML
TCA0833-025MLEA 135.27 USD
TCA0833-025ML TCA0833-500ML
1-(3-Aminopropyl)-2-methylpiperidine ≥98.0% (by GC, titration analysis)
1-(3-Aminopropyl)-2-methylpiperidine

CAS Number: 25560-00-3
MDL Number: MFCD00006517
Molecular Formula: C9H20N2
Molecular Weight: 156.27
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 170
Flash Point (°C): 88
Specific Gravity (20/20): 0.90


Other Product Names / Synonyms: 1-(3-Aminopropyl)-2-pipecoline

Formula: C₉H₂₀N₂
MW: 156.27 g/mol
Boiling Pt: 170 °C
Density: 0.90 (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD00006517
CAS Number: 25560-00-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d68b9774b19ec16277ae0a950ff222da5386b743&vid=d68b9774b19ec16277ae0a950ff222da5386b743&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR