Ethyl DL-3-acetoxybutyrate ≥98.0%
Supplier: TCI America
Synonyms:
DL-3-Acetoxybutyric Acid Ethyl Ester
A0831-5ML
A0831-25ML
TCA0831-005MLEA
29.51
USD
TCA0831-005ML
TCA0831-025ML
Ethyl DL-3-acetoxybutyrate ≥98.0%
Ethyl DL-3-acetoxybutyrate
CAS Number: 27846-49-7
MDL Number: MFCD00059378
Molecular Formula: C8H14O4
Molecular Weight: 174.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 94
Specific Gravity (20/20): 1.03
Other Product Names / Synonyms: DL-3-Acetoxybutyric Acid Ethyl Ester
Formula:
C₈H₁₄O₄ MW: 174.20 g/mol Boiling Pt: 94 °C Density: 1.03 (20 °C) |
MDL Number:
MFCD00059378 CAS Number: 27846-49-7 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...