3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepine ≥98.0%

Supplier: TCI America

Synonyms: 4-Chloro-2,2'-iminodibenzyl

C1866-25G C1866-5G
TCC1866-25GEA 139 USD
TCC1866-25G TCC1866-5G
3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepine ≥98.0%
3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepine

CAS Number: 32943-25-2
MDL Number: MFCD01632171
Molecular Formula: C14H12ClN
Molecular Weight: 229.71
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 87


Other Product Names / Synonyms: 4-Chloro-2,2'-iminodibenzyl

Formula: C₁₄H₁₂ClN
MW: 229.71 g/mol
Melting Pt: 87 °C
MDL Number: MFCD01632171
CAS Number: 32943-25-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b97c94eab1c9925032ac333791215e09de64116&vid=9b97c94eab1c9925032ac333791215e09de64116&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR