cis-6-Chloro-2-hexene ≥90.0%
Supplier: TCI America
Synonyms:
cis-4-Hexen-1-yl Chloride
C1653-5ML
C1653-25ML
TCC1653-005MLEA
192.5
USD
TCC1653-005ML
TCC1653-25ML
cis-6-Chloro-2-hexene ≥90.0%
cis-6-Chloro-2-hexene
CAS Number: 62614-70-4
MDL Number: MFCD02093458
Molecular Formula: C6H11Cl
Molecular Weight: 118.60
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 132
Specific Gravity (20/20): 0.92
Other Product Names / Synonyms: cis-4-Hexen-1-yl Chloride
Formula:
C₆H₁₁Cl MW: 118.60 g/mol Boiling Pt: 132 °C Density: 0.92 (20 °C) |
MDL Number:
MFCD02093458 CAS Number: 62614-70-4 |
Specification Test Results
Purity/Analysis Method | >90.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...