α-Chloropropiophenone ≥95.0%

Supplier: TCI America

C1647-25G C1647-5G
TCC1647-25GEA 485 USD
TCC1647-25G TCC1647-5G
α-Chloropropiophenone ≥95.0%
α-Chloropropiophenone

CAS Number: 6084-17-9
MDL Number: MFCD03844782
Molecular Formula: C9H9ClO
Molecular Weight: 168.62
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.16


Other Product Names / Synonyms: 2-Chloro-1-phenyl-1-propanone

Formula: C₉H₉ClO
Density: 1.16 (20 °C)
MDL Number: MFCD03844782
CAS Number: 6084-17-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4741c825e169780e2b16826db4beb81e55415519&vid=4741c825e169780e2b16826db4beb81e55415519&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR